Learn More
Selleck Chemical LLC trans-Zeatin-riboside
SDP
Catalog No. 5000003060
Encompass_Preferred
Zeatin Riboside is the most active and ubiquitous form of the naturally occurring cytokinins that promote cell division stimulate shoot proliferation inhibit root formation slow the aging process and activate gene expression and metabolic activity Zeatin riboside has an immunomodulatory effect by agonizing the mammalian adenosine A2A receptor
Catalog No. 50-000-03060 Supplier Selleck Chemical LLC Supplier No. S56785MG
Zeatin Riboside is the most active and ubiquitous form of the naturally occurring cytokinins that promote cell division stimulate shoot proliferation inhibit root formation slow the aging process and activate gene expression and metabolic activity Zeatin riboside has an immunomodulatory effect by agonizing the mammalian adenosine A2A receptor