Learn More
Medchemexpress LLC trans-Zeatinriboside | 6025-53-2 | 99.8% | 351.36 | 10 MM 1 ML
SDP
Catalog No. 5000452815
Encompass_Preferred
Trans-Zeatinriboside is a cytokinin precursor that plays a crucial role in plant development. It functions as a major long-distance signaling molecule within xylem vessels.
  • Acts as a major long-distance signaling form in xylem vessels.
  • Regulates leaf size and meristem activity-related traits.
Catalog No. 50-004-52815 Supplier Medchemexpress LLC Supplier No. HYW01115110MM/1ML
Trans-Zeatinriboside is a cytokinin precursor that plays a crucial role in plant development. It functions as a major long-distance signaling molecule within xylem vessels.
  • Acts as a major long-distance signaling form in xylem vessels.
  • Regulates leaf size and meristem activity-related traits.