Learn More
MSC Glove Guard Lp Glove Rings Accessory, Plastic, White, 12/Pk.
Catalog No. 5000685999
Encompass
The Glove Rings® accessory is designed to assist professionals in Hazmat services by reducing suit-up time while increasing wrist and hand mobility. Constructed from strong, flexible plastic, these rings provide a firm surface for taping, allowing for a better and more consistent seal.
  • Reduces suit-up time for increased efficiency in professional applications
  • Increases wrist and hand mobility
  • Provides a firm, stable surface for consistent taping and sealing
  • Features a micro-grip finish
  • Manufactured from strong, flexible plastic
  • Suitable for applications such as hydro-blasting
Catalog No. 50-006-85999 Supplier MSC Supplier No. 70954698
The Glove Rings® accessory is designed to assist professionals in Hazmat services by reducing suit-up time while increasing wrist and hand mobility. Constructed from strong, flexible plastic, these rings provide a firm surface for taping, allowing for a better and more consistent seal.
  • Reduces suit-up time for increased efficiency in professional applications
  • Increases wrist and hand mobility
  • Provides a firm, stable surface for consistent taping and sealing
  • Features a micro-grip finish
  • Manufactured from strong, flexible plastic
  • Suitable for applications such as hydro-blasting