Learn More
TARGETMOL CHEMICALS INC TRANSCAINIDE 5MG
SDP
Catalog No. 502580710
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Transcainide(R 54718) a new lidocaine analog is an orally active antiarrhythmic agent.Transcainide blocks the open state of BTX-activated sodium channels in bovine heart and rat skeletal muscle. purity: 100%
Catalog No. 50-258-0710 Supplier TARGETMOL CHEMICALS INC Supplier No. T262875MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Transcainide(R 54718) a new lidocaine analog is an orally active antiarrhythmic agent.Transcainide blocks the open state of BTX-activated sodium channels in bovine heart and rat skeletal muscle. purity: 100%