Learn More
Thomson Instrument Company Bidirectional Transfer Cap, 2' Tubing w/ Luer Lock, Sterile
SDP
Catalog No. 501120013
Encompass_Preferred
Thomson’s Bidirectional Transfer Caps enable precise liquid handling in and out of bioreactors, cell bags, and Optimum Growth flasks—compatible with peristaltic pump setup. Designed for high-throughput facilities, these caps support flexible media exchange while maintaining a closed aseptic system.
• Bi-directional design with luer locks, tubing, or quick connects enables aseptic media transfer, inoculation, or sampling into or out of 1.6L, 2.8L, 5L, and 7L flasks
• Quick to assemble, reducing process downtime
• Sterile, closed-loop design with multiple connection options enables seamless integration into custom systems while maintaining aseptic integrity
• Flexible setup—compatible with common fluid transfer systems and pumps
• Made in the USA and individually packaged for cleanroom use
Catalog No. 50-112-0013 Supplier Thomson Instrument Company Supplier No. 9318048
Add to Cart
Add to Cart
Thomson’s Bidirectional Transfer Caps enable precise liquid handling in and out of bioreactors, cell bags, and Optimum Growth flasks—compatible with peristaltic pump setup. Designed for high-throughput facilities, these caps support flexible media exchange while maintaining a closed aseptic system.
• Bi-directional design with luer locks, tubing, or quick connects enables aseptic media transfer, inoculation, or sampling into or out of 1.6L, 2.8L, 5L, and 7L flasks
• Quick to assemble, reducing process downtime
• Sterile, closed-loop design with multiple connection options enables seamless integration into custom systems while maintaining aseptic integrity
• Flexible setup—compatible with common fluid transfer systems and pumps
• Made in the USA and individually packaged for cleanroom use