Learn More
Bellbrook Labs Transcreener dAMP Exonuclease FP Assay
SDP
Catalog No. NC2951872
Encompass
The Transcreener dAMP Exonuclease FP Assay is a biochemical activity assay that measures the dAMP produced by exonuclease enzymes. This kit contains detection reagents for 10,000 assay wells in a 384-well format. The assay is designed for HTS, Hit-to-Lead, SAR, and general discovery and can be used with any plate reader that measures fluorescence polarization.
Catalog No. NC2951872 Supplier Bellbrook Labs Supplier No. 302810K
The Transcreener dAMP Exonuclease FP Assay is a biochemical activity assay that measures the dAMP produced by exonuclease enzymes. This kit contains detection reagents for 10,000 assay wells in a 384-well format. The assay is designed for HTS, Hit-to-Lead, SAR, and general discovery and can be used with any plate reader that measures fluorescence polarization.