Learn More
Sigma Aldrich Fine Chemicals Biosciences TranscreenerR UDP2 FP Assa10k
Catalog No. 502908538
Encompass
The Transcreener(R) UDP2 FP Assay is a universal biochemical HTS assay for enzymes that produce UDP including glycosyltransferase galactosyltransferase glucuronyltransferase N-acetylglucosamyltransferase N-acetlygalactosyltransferase xylosyltransferase and glycogen cellulose lactose and hyaluronan synthases. Enzyme activity is signaled by a decrease in fluorescence polarization as the bound tracer is displaced from the Transcreener(R) UDP2 Antibody. The assay is a simple single step mix-and-read format enabling the use of unmodified native substrate concentrations of 1 ndash 1000 muM. The assay provides excellent signal at low substrate conversion with a Zrsquo 0.7 and 100 millipolarization shift (mP) under normal reaction conditions.View full Transcreener(R) product list
Catalog No. 50-290-8538 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 301810K
Add to Cart
Add to Cart
Request a Quote
The Transcreener(R) UDP2 FP Assay is a universal biochemical HTS assay for enzymes that produce UDP including glycosyltransferase galactosyltransferase glucuronyltransferase N-acetylglucosamyltransferase N-acetlygalactosyltransferase xylosyltransferase and glycogen cellulose lactose and hyaluronan synthases. Enzyme activity is signaled by a decrease in fluorescence polarization as the bound tracer is displaced from the Transcreener(R) UDP2 Antibody. The assay is a simple single step mix-and-read format enabling the use of unmodified native substrate concentrations of 1 ndash 1000 muM. The assay provides excellent signal at low substrate conversion with a Zrsquo 0.7 and 100 millipolarization shift (mP) under normal reaction conditions.View full Transcreener(R) product list