Learn More
Sigma Aldrich Fine Chemicals Biosciences Transcript RNA Markers 0.2-10 kb, 50 uG
Catalog No. NC2004828
Encompass
Suitable for size determination of small RNA using native or denaturing agarose gel electrophoresis. Transcript RNA Markers 0.2-10 kb has been used as a molecular weight marker for northern blot hybridization.
Catalog No. NC2004828 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. R702050UG
Suitable for size determination of small RNA using native or denaturing agarose gel electrophoresis. Transcript RNA Markers 0.2-10 kb has been used as a molecular weight marker for northern blot hybridization.