Learn More
Cusabio Technology LLC Transcription activator BRG1
Catalog No. 5000051049
Encompass_Preferred
This kit detects SMARCA4 (BRG1) in human serum plasma tissue homogenates and cell lysates using a sandwich ELISA technique It offers a quantitative detection range of 25 1600 pg/mL with a sensitivity of 6 25 pg/mL enabling precise measurement of low-abundance targets The assay requires 50 100 L of sample and delivers results within 1 5 hours Optical density is measured at 450 nm to determine SMARCA4 concentrations validated for research or diagnostic applications requiring reliable protein quantification Its robust design ensures reproducibility across diverse biological matrices while maintaining high specificity for the transcription activator BRG1
Catalog No. 50-000-51049 Supplier Cusabio Technology LLC Supplier No. CSBEL021801HU
This kit detects SMARCA4 (BRG1) in human serum plasma tissue homogenates and cell lysates using a sandwich ELISA technique It offers a quantitative detection range of 25 1600 pg/mL with a sensitivity of 6 25 pg/mL enabling precise measurement of low-abundance targets The assay requires 50 100 L of sample and delivers results within 1 5 hours Optical density is measured at 450 nm to determine SMARCA4 concentrations validated for research or diagnostic applications requiring reliable protein quantification Its robust design ensures reproducibility across diverse biological matrices while maintaining high specificity for the transcription activator BRG1