Learn More
Roche Diagnostics TRANSCRIPTOR 1ST STRD CDNA KIT
Supplier:  Roche Diagnostics 04379012001
Encompass
Transcriptor First Strand cDNA Synthesis Kit; Contains a recombinant reverse transcriptase for the robust transcription of RNA fragments up to 14 kb
Catalog No. 50-997-721