Learn More
Sigma Aldrich Fine Chemicals Biosciences Transcriptor One Step RT P
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 4655885001
Encompass
The Transcriptor One-Step RT-PCR Kit provides all components required for one-step RT-PCR (except primers and template) in a convenient format. The 50-reaction pack size also includes 10 control reactions (control RNA and control primer mix).The kits RT-PCR Enzyme Mix contains four different enzymes Transcriptor Reverse Transcriptase ensures sensitive and robust reverse transcription with high yield Protector RNase Inhibitor fully active at elevated temperatures provides maximum template protection during reverse transcription and the Expand System - a blend consisting of Taq DNA Polymerase and a proofreading polymerase - minimizes the possibility of mutations offering high yield and fidelity in the PCR.The optimized RT-PCR Reaction Buffer includes high quality dNTPs and provides the overall improved performance of a hot start system. The buffers unique hot start component binds and sequesters primers at lower temperatures to prevent the primers from binding to nonspecific sites. An
Catalog No. 50-291-4140