Learn More
TARGETMOL CHEMICALS INC Transcrocetinate disodium 10MG
SDP
Catalog No. 502880426
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Transcrocetinate disodium (Disodium trans-crocetinate), extracted from saffron (Crocus sativus L.), is a high-affinity antagonist of NMDA receptor. Purity 99.22%
Catalog No. 50-288-0426 Supplier TARGETMOL CHEMICALS INC Supplier No. TQ002710MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Transcrocetinate disodium (Disodium trans-crocetinate), extracted from saffron (Crocus sativus L.), is a high-affinity antagonist of NMDA receptor. Purity 99.22%