Learn More
Waters Corp Waters Pressure Transducer, 20K PSI, UPLC System Compatible
Catalog No. 50201699 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement pressure transducer is designed for use in various ACQUITY systems, ensuring precise pressure measurement and maintaining system performance.
  • Functions as a replacement pressure transducer
  • Compatible with ACQUITY I-Class Binary Solvent Managers
  • Compatible with ACQUITY M-Class Binary and Auxiliary Solvent Managers
  • Compatible with ACQUITY Isocratic Solvent
  • Designed for UPLC system types
  • Rated for 20,000 PSI pressure
  • Comes with 1 unit per package
Catalog No. 50-201-699 Supplier Waters Corp Supplier No. 700006045
This replacement pressure transducer is designed for use in various ACQUITY systems, ensuring precise pressure measurement and maintaining system performance.
  • Functions as a replacement pressure transducer
  • Compatible with ACQUITY I-Class Binary Solvent Managers
  • Compatible with ACQUITY M-Class Binary and Auxiliary Solvent Managers
  • Compatible with ACQUITY Isocratic Solvent
  • Designed for UPLC system types
  • Rated for 20,000 PSI pressure
  • Comes with 1 unit per package