Learn More
Waters Corp Waters Corporation Transducer Assembly, Head Mounted, 15KPSI, UPLC System
Catalog No. 50792771 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This pressure transducer assembly is designed as a replacement component for UPLC systems. It mounts directly onto the pump head and is compatible with ACQUITY and nanoACQUITY solvent managers.
  • Designed for head-mounted installation on pump heads.
  • Provides 15KPSI pressure capability.
  • Suitable for UPLC systems.
  • Utilized on compatible solvent managers.
Catalog No. 50-792-771 Supplier Waters Corp Supplier No. 700002594
This pressure transducer assembly is designed as a replacement component for UPLC systems. It mounts directly onto the pump head and is compatible with ACQUITY and nanoACQUITY solvent managers.
  • Designed for head-mounted installation on pump heads.
  • Provides 15KPSI pressure capability.
  • Suitable for UPLC systems.
  • Utilized on compatible solvent managers.