Learn More
System Biosciences LLC TRANSDUX MAX LENTIVIRUS TRANSDUCTION REAGENTTransDux MAX Lentivirus Transduction Enhancer = 100 Transductions
SDP
Catalog No. NC1316348
Encompass_Preferred
Whether you’re working with difficult-to-transduce cells like primary T-cells or just want to ensure high transduction efficiency, SBI’s TransDux™ reagents are ready to deliver. Our newer TransDux MAX Lentiviral Transduction Reagent can increase transduction efficiencies by up to 8-fold compared to polybrene, while our original TransDux formulation (Cat# LV850A-1) is still available for researchers who are not yet ready to make the change.Ensure reliable and efficient gene delivery with TransDux MAX.
Catalog No. NC1316348 Supplier System Biosciences LLC Supplier No. LV860A1
Whether you’re working with difficult-to-transduce cells like primary T-cells or just want to ensure high transduction efficiency, SBI’s TransDux™ reagents are ready to deliver. Our newer TransDux MAX Lentiviral Transduction Reagent can increase transduction efficiencies by up to 8-fold compared to polybrene, while our original TransDux formulation (Cat# LV850A-1) is still available for researchers who are not yet ready to make the change.Ensure reliable and efficient gene delivery with TransDux MAX.