Learn More
Bio Rad Laboratories TRANSFECTIN LIPID REAGENT
Catalog No. 509012643
Encompass
Lipid Reagent; Bio-Rad; TransFectin; High efficiency and gene activity; Transfection at 50 to 90% culture confluence; Simple three-step protocol; Affordable; Storage: 4 deg.C; 1.0mL; Quantity: 5
Catalog No. 50-901-2643 Supplier Bio Rad Laboratories Supplier No. 1703352
Lipid Reagent; Bio-Rad; TransFectin; High efficiency and gene activity; Transfection at 50 to 90% culture confluence; Simple three-step protocol; Affordable; Storage: 4 deg.C; 1.0mL; Quantity: 5