Learn More
Macherey-Nagel NucleoSpin 96 Plasmid Transfection-grade Core Kit, 4 x 96 preps
Catalog No. NC1188502
Encompass_Preferred
NucleoSpin 96 Plasmid Transfection-grade Core Kit (4 x 96) 4 x 96 preps for the isolation of plasmid DNA - NucleoSpin Filter Plates, NucleoSpin Plasmid Binding Plates, buffers, RNase A
Catalog No. NC1188502 Supplier Macherey-Nagel Supplier No. 740492.4
NucleoSpin 96 Plasmid Transfection-grade Core Kit (4 x 96) 4 x 96 preps for the isolation of plasmid DNA - NucleoSpin Filter Plates, NucleoSpin Plasmid Binding Plates, buffers, RNase A