Learn More
Lonza Walkersville Inspect Equip 4D-Nufe 1 Cuvett
Catalog No. 5000496477
Encompass
The inspection equipment is a fixed resistance version of the 100 microL Nucleocuvette Vessel It can be used to check basic functionality of the 4D-Nucleofector X Unit prior to use with single cuvettes
Catalog No. 50-004-96477 Supplier Lonza Walkersville Supplier No. AXP1003
The inspection equipment is a fixed resistance version of the 100 microL Nucleocuvette Vessel It can be used to check basic functionality of the 4D-Nucleofector X Unit prior to use with single cuvettes