Learn More
Lonza Walkersville Inspt Equip 4D-Nufe 16wl Strip
Catalog No. 5000496476
Encompass
The inspection equipment is a fixed resistance version of the 16-well Nucleocuvette Strip It can be used to check basic functionality of the 4D-Nucleofector X Unit prior to use with the strips
Catalog No. 50-004-96476 Supplier Lonza Walkersville Supplier No. AXP1004
The inspection equipment is a fixed resistance version of the 16-well Nucleocuvette Strip It can be used to check basic functionality of the 4D-Nucleofector X Unit prior to use with the strips