Learn More
Lonza Walkersville Inspect Equip 96-well Shuttle
Catalog No. 5000496474
Encompass
The inspection equipment is a fixed resistance version of the 96-well Nucleocuvette Plate that can be used to check basic functionality of the 96-well Shuttle Add-On prior to use
Catalog No. 50-004-96474 Supplier Lonza Walkersville Supplier No. AXP1002
The inspection equipment is a fixed resistance version of the 96-well Nucleocuvette Plate that can be used to check basic functionality of the 96-well Shuttle Add-On prior to use