Learn More
Lonza Walkersville Inspect Equipment Nufe II/2b
Catalog No. 5000496475
Encompass
The inspection equipment is a fixed resistance version of the 100 L cuvette It can be used to check basic functionality of the Nucleofector 2b System (or older generations I or II) prior to use
Catalog No. 50-004-96475 Supplier Lonza Walkersville Supplier No. AXP1001
The inspection equipment is a fixed resistance version of the 100 L cuvette It can be used to check basic functionality of the Nucleofector 2b System (or older generations I or II) prior to use