Learn More
EDITCO BIO INC Transfection Optimization Kit (Multi-guide)
Catalog No. NC1842432
Encompass
The Transfection Optimization Kit is a validated positive control kit based on Synthego’s multi-guide strategy that targets human TRAC gene. Recommend using this kit to optimize transfection conditions for your cell type prior to using the Gene Knockout Kit v2. Consider a cell line has been optimized when you achieve at least 50% editing efficiency for TRAC positive control.
Catalog No. NC1842432 Supplier EDITCO BIO INC Supplier No. TOK_MULTIGUIDE_00000
The Transfection Optimization Kit is a validated positive control kit based on Synthego’s multi-guide strategy that targets human TRAC gene. Recommend using this kit to optimize transfection conditions for your cell type prior to using the Gene Knockout Kit v2. Consider a cell line has been optimized when you achieve at least 50% editing efficiency for TRAC positive control.