Learn More
System Biosciences LLC Cas9 SmartNuclease™ mRNA, Injection- and Transfection-readyTransfection-ready Cas9 SmartNuclease mRNA (Eukaryotic version) = 20ug
SDP
Catalog No. NC0561970
Encompass_Preferred
From basic biological studies to generating novel disease models, SBI’s injection- and transfection-ready PrecisonX™ Cas9 SmartNuclease™ mRNA is here to move your in vivo genome engineering projects forward faster.Use with an appropriate gRNA and, if your application needs it, a homologous recombination (HR) targeting vector for gene knock-ins, knock-outs, editing, and tagging. The Cas9 SmartNuclease will generate a double-strand break (DSB) at the site specified by the gRNA, and this DSB can be efficiently and precisely repaired in a manner that includes your desired mutation through the use of an HR targeting vector.
Catalog No. NC0561970 Supplier System Biosciences LLC Supplier No. CAS500A1
From basic biological studies to generating novel disease models, SBI’s injection- and transfection-ready PrecisonX™ Cas9 SmartNuclease™ mRNA is here to move your in vivo genome engineering projects forward faster.Use with an appropriate gRNA and, if your application needs it, a homologous recombination (HR) targeting vector for gene knock-ins, knock-outs, editing, and tagging. The Cas9 SmartNuclease will generate a double-strand break (DSB) at the site specified by the gRNA, and this DSB can be efficiently and precisely repaired in a manner that includes your desired mutation through the use of an HR targeting vector.