Learn More
Andwin Scientific TRANSFECTION REAGENT + BUFFER
SDP
Catalog No. NC0340364
Encompass_Preferred
The jetPRIME™ DNA and siRNA Transfection Reagent is made to guarantee high DNA transfection efficiency and excellent gene silencing. This product is very cost-effective as it uses less reagent and DNA and provides more gentle transfections in comparison to other competitors. It can be used with no difficulty and with serum and antibiotics. This can produce 188 reactions in 6-well plates and has a buffer volume of 60 mL. It is sold individually.
Catalog No. NC0340364 Supplier Andwin Scientific Supplier No. 89129922
The jetPRIME™ DNA and siRNA Transfection Reagent is made to guarantee high DNA transfection efficiency and excellent gene silencing. This product is very cost-effective as it uses less reagent and DNA and provides more gentle transfections in comparison to other competitors. It can be used with no difficulty and with serum and antibiotics. This can produce 188 reactions in 6-well plates and has a buffer volume of 60 mL. It is sold individually.