Learn More
Lonza Walkersville 4D-Nucleofector Automat Pack
Catalog No. 5000496323
Encompass
The 4D-Nucleofector Automation Package provides the interface that is required for using the 4D-Nucleofector 96-well Unit in a liquid handling system for automated medium throughput Nucleofection It also includes an interface documentation and potential support for integration
Catalog No. 50-004-96323 Supplier Lonza Walkersville Supplier No. SBA3001
The 4D-Nucleofector Automation Package provides the interface that is required for using the 4D-Nucleofector 96-well Unit in a liquid handling system for automated medium throughput Nucleofection It also includes an interface documentation and potential support for integration