Learn More
Lonza Walkersville 4D-Nucleofector LogWare
Catalog No. 5000496460
Encompass
For the 4D-Nucleofector System Lonza offers accessory products which provide higher quality standards for transfection applications in upstream GMP manufacturing environments The 4D-Nucleofector LogWare is a 21CFR part 11 compliant software version of the standard software that supports the use of the 4D-Nucleofector System in a GMP environment A validation documentation is available on request
Catalog No. 50-004-96460 Supplier Lonza Walkersville Supplier No. SAAF1001
For the 4D-Nucleofector System Lonza offers accessory products which provide higher quality standards for transfection applications in upstream GMP manufacturing environments The 4D-Nucleofector LogWare is a 21CFR part 11 compliant software version of the standard software that supports the use of the 4D-Nucleofector System in a GMP environment A validation documentation is available on request