Learn More
Lonza Walkersville 4D-Nucleofector LV Reservoir
Supplier:  Lonza Walkersville V4LR1001W
Encompass
The 4D-Nucleofector LV Reservoirs were developed as optimal input tubes and collection tubes used in conjunction with the flow-through LV Nucleocuvette Cartridge when using the 4D-Nucleofector LV Unit They are used to load the 4D-Nucleofector LV Unit and to collect the transfected cells Two of such 4D-Nucleofector LV Reservoirs are provided with each 4D-Nucleofector LV Kit XL which contains one LV Nucleocuvette Cartridge This additional 4D-Nucleofector LV Reservoir is required when the second input line of an LV Nucleocuvette Cartridge shall be used for separate feeding of mRNA
Catalog No. 50-004-96459