Learn More
Lonza Walkersville Mouse Emb Fibro Start Nufe Kit
Catalog No. 5000496443
Encompass
The Nucleofector 2b System (or older generations I or II) works cell type specific kits each of them dedicated to an individual primary cell Due to significant diversity between different MEF clones we have developed two Nucleofector Kits (1 and 2) and an easy-to-use Nucleofector Starter Kit The Mouse Embryonic Fibroblast Nucleofector Starter Kit is used to determine which of the two Nucleofector Kits is best suited for transfecting a specific primary mouse embyonic fibroblast (MEF) clone in Nucleofector 2b System The respective cell type-specific Optimized Protocol or recommendation is available under Instructions or in our Lonza Bioscience knowledge database In this Optimized Protocol the best Nucleofection conditions are indicated In addition we share our experience and knowledge for treatment of this cell type
Catalog No. 50-004-96443 Supplier Lonza Walkersville Supplier No. VPD1006
The Nucleofector 2b System (or older generations I or II) works cell type specific kits each of them dedicated to an individual primary cell Due to significant diversity between different MEF clones we have developed two Nucleofector Kits (1 and 2) and an easy-to-use Nucleofector Starter Kit The Mouse Embryonic Fibroblast Nucleofector Starter Kit is used to determine which of the two Nucleofector Kits is best suited for transfecting a specific primary mouse embyonic fibroblast (MEF) clone in Nucleofector 2b System The respective cell type-specific Optimized Protocol or recommendation is available under Instructions or in our Lonza Bioscience knowledge database In this Optimized Protocol the best Nucleofection conditions are indicated In addition we share our experience and knowledge for treatment of this cell type