Learn More
Lonza Walkersville P3 Prim Cell 384-well Nufe Kit
Catalog No. 5000496367
Encompass
The P3 Primary Cell 384-well Nucleofector Kit is one of our five kits suited for high-throughput transfection of primary cells when working with the 384-well HT Nucleofector System This kit has been determined to be optimal for transfecting following cell types B cell (human) CD34 cell (human) Chondrocyte (human) DC (human) DC (mouse) mature ES (human) H9 ES (mouse) Hepatocyte (human) HMEC (human) Keratinocyte (human) Macrophage (human) Monocyte (human) Neuron (rat) brain Hi/Cx NHBE (human) RPE (human) T cell (human) stim T cell (mouse) unstim The respective cell type-specific Optimized Protocols or recommendations are available under Instructions or in our Lonza Bioscience knowledge database In these Optimized Protocols the best Nucleofection conditions are indicated In addition we share our experience and knowledge for treatment of individual cell types The kit is available in different variants for either 2 x 384 or 10 x 384 reactions
Catalog No. 50-004-96367 Supplier Lonza Walkersville Supplier No. V5SP3002
The P3 Primary Cell 384-well Nucleofector Kit is one of our five kits suited for high-throughput transfection of primary cells when working with the 384-well HT Nucleofector System This kit has been determined to be optimal for transfecting following cell types B cell (human) CD34 cell (human) Chondrocyte (human) DC (human) DC (mouse) mature ES (human) H9 ES (mouse) Hepatocyte (human) HMEC (human) Keratinocyte (human) Macrophage (human) Monocyte (human) Neuron (rat) brain Hi/Cx NHBE (human) RPE (human) T cell (human) stim T cell (mouse) unstim The respective cell type-specific Optimized Protocols or recommendations are available under Instructions or in our Lonza Bioscience knowledge database In these Optimized Protocols the best Nucleofection conditions are indicated In addition we share our experience and knowledge for treatment of individual cell types The kit is available in different variants for either 2 x 384 or 10 x 384 reactions