Learn More
Lonza Walkersville Mouse T Cell Nucleofector Kit
Catalog No. 5000496426
Encompass
The Nucleofector 2b System (or older generations I or II) works cell type specific kits each of them dedicated to an individual primary cell This Nucleofector Kit is the optimal kit for efficient transfection of primary mouse T cells The respective cell type-specific Optimized Protocol or recommendation is available under Instructions or in our Lonza Bioscience knowledge database In this Optimized Protocol the best Nucleofection conditions are indicated In addition we share our experience and knowledge for treatment of this cell type
Catalog No. 50-004-96426 Supplier Lonza Walkersville Supplier No. VPA1006
The Nucleofector 2b System (or older generations I or II) works cell type specific kits each of them dedicated to an individual primary cell This Nucleofector Kit is the optimal kit for efficient transfection of primary mouse T cells The respective cell type-specific Optimized Protocol or recommendation is available under Instructions or in our Lonza Bioscience knowledge database In this Optimized Protocol the best Nucleofection conditions are indicated In addition we share our experience and knowledge for treatment of this cell type