Learn More
Lonza Walkersville P4 PC 384-well Nufe Kit L
Catalog No. 5000496431
Encompass
The P4 Primary Cell 384-well Nucleofector Kit is one of our five kits suited for high-throughput transfection of primary cells when working with the 384-well HT Nucleofector System This kit has been determined to be optimal for transfecting following cell types B cell (mouse) DC (mouse) immat MEF (mouse) The respective cell type-specific Optimized Protocols or recommendations are available under Instructions or in our Lonza Bioscience knowledge database In these Optimized Protocols the best Nucleofection conditions are indicated In addition we share our experience and knowledge for treatment of individual cell types The kit is available in different variants for either 2 x 384 or 10 x 384 reactions
Catalog No. 50-004-96431 Supplier Lonza Walkersville Supplier No. V5SP4010
The P4 Primary Cell 384-well Nucleofector Kit is one of our five kits suited for high-throughput transfection of primary cells when working with the 384-well HT Nucleofector System This kit has been determined to be optimal for transfecting following cell types B cell (mouse) DC (mouse) immat MEF (mouse) The respective cell type-specific Optimized Protocols or recommendations are available under Instructions or in our Lonza Bioscience knowledge database In these Optimized Protocols the best Nucleofection conditions are indicated In addition we share our experience and knowledge for treatment of individual cell types The kit is available in different variants for either 2 x 384 or 10 x 384 reactions