Learn More
Lonza Walkersville Hu Dendritic Cell Nufe Kit
Catalog No. 5000496473
Encompass
The Nucleofector 2b System (or older generations I or II) works cell type specific kits each of them dedicated to an individual primary cell This Nucleofector Kit is the optimal kit for efficient transfection of primary human dendritic cells (immature or mature) The respective cell type-specific Optimized Protocol or recommendation is available under Instructions or in our Lonza Bioscience knowledge database In this Optimized Protocol the best Nucleofection conditions are indicated In addition we share our experience and knowledge for treatment of this cell type
Catalog No. 50-004-96473 Supplier Lonza Walkersville Supplier No. VPA1004
The Nucleofector 2b System (or older generations I or II) works cell type specific kits each of them dedicated to an individual primary cell This Nucleofector Kit is the optimal kit for efficient transfection of primary human dendritic cells (immature or mature) The respective cell type-specific Optimized Protocol or recommendation is available under Instructions or in our Lonza Bioscience knowledge database In this Optimized Protocol the best Nucleofection conditions are indicated In addition we share our experience and knowledge for treatment of this cell type