Learn More
Lonza Walkersville Prim Cell Opt 4D-Nufe Y Kit
Catalog No. 5000496428
Encompass
Electroporation-based methods have so far required cells to be in suspension for transfection However the 4D-Nucleofector Y Unit allows direct Nucleofection of cells in adherence Cells which typically grow in adherence in culture can be kept and transfected by Nucleofection in their physiological state The Primary Cell Optimization 4D-Nucleofector Y Kit is suited for rapid determine Nucleofection conditions for primary cell types lacking an Optimized Protocol for the 4D-Nucleofector Y Unit Using one 24-well Dipping Electrode Array and our pmaxGFP Vector as positive control our two Primary Cell Nucleofector Solutions for adherent Nucleofection (AD1 and AD2) are tested in combination with a pre-selected set of programs plus controls The remaining 24-well Dipping Electrode Array can be used for optional fine tuning or performing the first experiment with the selected conditions
Catalog No. 50-004-96428 Supplier Lonza Walkersville Supplier No. V4YP9A48
Electroporation-based methods have so far required cells to be in suspension for transfection However the 4D-Nucleofector Y Unit allows direct Nucleofection of cells in adherence Cells which typically grow in adherence in culture can be kept and transfected by Nucleofection in their physiological state The Primary Cell Optimization 4D-Nucleofector Y Kit is suited for rapid determine Nucleofection conditions for primary cell types lacking an Optimized Protocol for the 4D-Nucleofector Y Unit Using one 24-well Dipping Electrode Array and our pmaxGFP Vector as positive control our two Primary Cell Nucleofector Solutions for adherent Nucleofection (AD1 and AD2) are tested in combination with a pre-selected set of programs plus controls The remaining 24-well Dipping Electrode Array can be used for optional fine tuning or performing the first experiment with the selected conditions