Learn More
Lonza Walkersville P1 Prim Cell 96-well Nufe Kit
Catalog No. 5000496286
Encompass
The P1 Primary Cell 96-well Nucleofector Kit is one of our five kits suited for medium-throughput transfection of primary cells when working with the 4D-Nucleofector 96-well Unit This kit has been determined to be optimal for transfecting following cell types ADSC undiff (human) MSC (human) Preadipocytes (human) PrEC (human) The respective cell type-specific Optimized Protocols or recommendations are available under Instructions or in our Lonza Bioscience knowledge database In these Optimized Protocols the best Nucleofection conditions are indicated In addition we share our experience and knowledge for treatment of individual cell types The kit is available in different variants for either 1 x 96 or 10 x 96 reactions
Catalog No. 50-004-96286 Supplier Lonza Walkersville Supplier No. V4SP1960
The P1 Primary Cell 96-well Nucleofector Kit is one of our five kits suited for medium-throughput transfection of primary cells when working with the 4D-Nucleofector 96-well Unit This kit has been determined to be optimal for transfecting following cell types ADSC undiff (human) MSC (human) Preadipocytes (human) PrEC (human) The respective cell type-specific Optimized Protocols or recommendations are available under Instructions or in our Lonza Bioscience knowledge database In these Optimized Protocols the best Nucleofection conditions are indicated In addition we share our experience and knowledge for treatment of individual cell types The kit is available in different variants for either 1 x 96 or 10 x 96 reactions