Learn More
Lonza Walkersville AD2 4D-Nucleofector Y Kit
Supplier:  Lonza Walkersville V4YP2A24
Encompass
Electroporation-based methods have so far required cells to be in suspension for transfection However the 4D-Nucleofector Y Unit allows direct Nucleofection of cells in adherence Cells which typically grow in adherence in cell culture can be kept and transfected by Nucleofection in their physiological state The AD2 Primary Cell 4D-Nucleofector Y Kit is one of our two kits that are suited for electroporation of primary cells in adherence when working with the 4D-Nucleofector Y Unit The cells are cultured in the 24-well culture plates provided with the kit and for the transfection step a 24-well Dipping Electrode Array is inserted after replacing culture medium by the specific Nucleofector Solution mixed with the DNA or RNA substrate
Catalog No. 50-004-96254