Learn More
Lonza Walkersville Rat Cardiomyocyte Neo Nufe Kit
Catalog No. 5000496342
Encompass
The Nucleofector 2b System (or older generations I or II) works cell type specific kits each of them dedicated to an individual primary cell This Nucleofector Kit is the optimal kit for efficient transfection of primary rat cardiomocytes The respective cell type-specific Optimized Protocol or recommendation is available under Instructions or in our Lonza Bioscience knowledge database In this Optimized Protocol the best Nucleofection conditions are indicated In addition we share our experience and knowledge for treatment of this cell type The kit is available in different variants for either 10 or 25 or 4 x 25 reactions
Catalog No. 50-004-96342 Supplier Lonza Walkersville Supplier No. VPE1002
The Nucleofector 2b System (or older generations I or II) works cell type specific kits each of them dedicated to an individual primary cell This Nucleofector Kit is the optimal kit for efficient transfection of primary rat cardiomocytes The respective cell type-specific Optimized Protocol or recommendation is available under Instructions or in our Lonza Bioscience knowledge database In this Optimized Protocol the best Nucleofection conditions are indicated In addition we share our experience and knowledge for treatment of this cell type The kit is available in different variants for either 10 or 25 or 4 x 25 reactions