Learn More
FUJIFILM Biosciences Transfectory Supplement
Catalog No. NC1203712
Encompass
Part of the BalanCD Transfectory CHO system of media and supplements, the Transfectory Supplement is an animal component-free concentrated hydrolysate supplement that can be used to enhance cell growth and protein expression in CHO and various cell types.
Catalog No. NC1203712 Supplier FUJIFILM Biosciences Supplier No. 91148500ML
Part of the BalanCD Transfectory CHO system of media and supplements, the Transfectory Supplement is an animal component-free concentrated hydrolysate supplement that can be used to enhance cell growth and protein expression in CHO and various cell types.