Learn More
Chemglass Life Sciences Transfer Assembly, 3L, with Plugged End and Weldable C-Flex Tubing, Sterile
SDP
Supplier:  Chemglass Life Sciences CLS34653LA
Encompass_Preferred
Transfer cap assemblies for 3LFernbach flasks.The venting port ends in a 50mm diameter vent filter with a 0. 20um pore PTFE (hydrophobic) membrane and barbed fitting.On the external side you will find 1/8" ID x 1/4" OD weldable C-Flex tubing with a tubing clamp and ending with a 1/8" end tube plug.The liquid transfer port has an internal end with a rigid 1/8" x 1/4" OD C-Flex dip tube to reach the bottom of the flask.Individually double bagged and gamma irradiated sterile.
Catalog No. 50-000-83266