Learn More
Chemglass Life Sciences Transfer Assembly, 3L, with Plugged End and Weldable C-Flex Tubing, Sterile
SDP
Catalog No. NC1843194
Encompass_Preferred
Transfer cap assemblies for 3L Corning Fernbach flasks.The venting port ends in a 50mm diameter vent filter with a 0. 20um pore PTFE (hydrophobic) membrane and barbed fitting.On the external side you will find 1/8" ID x 1/4" OD weldable C-Flex tubing with a tubing clamp and ending with a 1/8" end tube plug.The liquid transfer port has an internal end with a rigid 1/8" x 1/4" OD C-Flex dip tube to reach the bottom of the flask.Individually double bagged and gamma irradiated sterile.
Catalog No. NC1843194 Supplier Chemglass Life Sciences Supplier No. CLS34653LC
Transfer cap assemblies for 3L Corning Fernbach flasks.The venting port ends in a 50mm diameter vent filter with a 0. 20um pore PTFE (hydrophobic) membrane and barbed fitting.On the external side you will find 1/8" ID x 1/4" OD weldable C-Flex tubing with a tubing clamp and ending with a 1/8" end tube plug.The liquid transfer port has an internal end with a rigid 1/8" x 1/4" OD C-Flex dip tube to reach the bottom of the flask.Individually double bagged and gamma irradiated sterile.