Learn More
Chemglass Life Sciences Transfer Cap Assembly, 125mL, Advantaflex Tubing, 38-430, Vented, Plugged End, Sterile
SDP
Catalog No. NC3720096
Encompass
Transfer cap assemblies, vented for Single Use Media bottles are a cost effective and convenient way to transfer liquid.The venting port ends in a 30mm diameter vent filter with a 0. 20um pore PTFE (hydrophobic) membrane and barbed fitting.All components in contact with the media are USP Class VI and the Size 16 tubing has a clamp and a plugged end.Tubing with a tubing clamp and ending with a plugged end.Individually double bagged and gamma irradiated sterile.
Catalog No. NC3720096 Supplier Chemglass Life Sciences Supplier No. CLS11610125
Transfer cap assemblies, vented for Single Use Media bottles are a cost effective and convenient way to transfer liquid.The venting port ends in a 30mm diameter vent filter with a 0. 20um pore PTFE (hydrophobic) membrane and barbed fitting.All components in contact with the media are USP Class VI and the Size 16 tubing has a clamp and a plugged end.Tubing with a tubing clamp and ending with a plugged end.Individually double bagged and gamma irradiated sterile.