Learn More
Chemglass Life Sciences Transfer Cap Assembly, 2, 000mL, Advantaflex Tubing, 53B, Vented, Plugged End, Sterile
SDP
Catalog No. NC3720088
Encompass
Transfer cap assemblies, vented for Single Use Media bottles are a cost effective and convenient way to transfer liquid.The venting port ends in a 30mm diameter vent filter with a 0. 20um pore PTFE (hydrophobic) membrane and barbed fitting.All components in contact with the media are USP Class VI and the Size 16 tubing has a clamp and a plugged end.Tubing with a tubing clamp and ending with a plugged end.Individually double bagged and gamma irradiated sterile.
Catalog No. NC3720088 Supplier Chemglass Life Sciences Supplier No. CLS11612000
Transfer cap assemblies, vented for Single Use Media bottles are a cost effective and convenient way to transfer liquid.The venting port ends in a 30mm diameter vent filter with a 0. 20um pore PTFE (hydrophobic) membrane and barbed fitting.All components in contact with the media are USP Class VI and the Size 16 tubing has a clamp and a plugged end.Tubing with a tubing clamp and ending with a plugged end.Individually double bagged and gamma irradiated sterile.