Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITL BioMedical Transfer Cap Set

Catalog No. 22799905 Shop All ITL BioMedical Products
Change view
Click to view available options
Quantity:
30/Pk., 6 Pk./Cs., 180/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-799-905 30/Pk., 6 Pk./Cs., 180/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-799-905 Supplier ITL BioMedical Supplier No. A100730
Add to Cart
Add to Cart

Safely and easily transfer sample from blood culture bottle to vacuum tubes. Perfect for labs using automated platforms.

  • All-in-one needleless blood culture sample transfer solution
  • Integrated safety lid
  • Plastic safety tip replaces needle for piercing culture bottle stopper
  • Flexible fit with most blood culture bottles and blood sample tubes
  • Contained sample transfer reduces potential exposure to biohazards
  • Applications include specimen sampling for automated equipment, sample splitting, retention samples
  • Compliance

    CE marked

    Specifications

    Sterility Sterile
    Color Clear
    Description Single use only
    For Use With (Application) Blood Culture
    For Use With (Equipment) BD Kiestra, Copan Wasp
    Packaging Individually packaged
    Quantity 30/Pk., 6 Pk./Cs., 180/Cs.
    Tip Material Plastic
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.