Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Transfer/Electro Blotting 10X Solution, 1 L
SDP

Catalog No. 501978311
Encompass_Preferred

Formulation:

TRIS Base: 0.25M
Glycine: 1.92M
pH: 8.5
Add Methanol according to protocol

Catalog No. 50-197-8311 Supplier Research Products International Corp Supplier No. T274501000.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Formulation:

TRIS Base: 0.25M
Glycine: 1.92M
pH: 8.5
Add Methanol according to protocol

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.