Learn More
Andwin Scientific TRANSFER PIPET 15.6 NS 500/PK
SDP
Catalog No. 50727604
Encompass_Preferred
The General Purpose Transfer Pipet have several uses, including but not limited to blood banking, reagent handling, and viscous liquid sampling and transferring. This product has an attached bulb that eliminates the risk of cross-contamination. It also features a low-affinity surface. This is sold in a pack of 500.
Catalog No. 50-727-604 Supplier Andwin Scientific Supplier No. 709601
The General Purpose Transfer Pipet have several uses, including but not limited to blood banking, reagent handling, and viscous liquid sampling and transferring. This product has an attached bulb that eliminates the risk of cross-contamination. It also features a low-affinity surface. This is sold in a pack of 500.