Learn More
BIOLOGIX USA INC 1ml Bulk/Individually Wrapped Transfer Pipets, Non Sterile, 500/case
Catalog No. NC3762190
Encompass_Preferred
1ml Bulk Transfer PipetsMaterial: PE, will not shatter or breakAn excellent alternative to Pasteur pipettes and rubber bulbsEvenly distributed thickness ensures uniform suction forceNon-sterilizedbulk package (non-sterile)DNase & RNase freeTransparent wall permits easy viewing of pipette contentsWith molded graduations on the body of the transfer pipettes
Catalog No. NC3762190 Supplier BIOLOGIX USA INC Supplier No. 300135
1ml Bulk Transfer PipetsMaterial: PE, will not shatter or breakAn excellent alternative to Pasteur pipettes and rubber bulbsEvenly distributed thickness ensures uniform suction forceNon-sterilizedbulk package (non-sterile)DNase & RNase freeTransparent wall permits easy viewing of pipette contentsWith molded graduations on the body of the transfer pipettes