Learn More
Alkali Scientific Transfer Pipette, 1.5mL Small Bulb Extended Tip, 20/pk, 500/cs
SDP
Catalog No. 502389527
Encompass_Preferred
These unbreakable, one-piece pipettes eliminate contaminated rubber bulbs, broken glass, and exposure to infectious materials. They are made of ultra-clear, low density polyethylene (LDPE) and are ideal for microbiology, blood banking, hematology, immunology, and urinalysis. Their low-affinity surface reduces sample, cell, or protein loss, and they can also be heat-sealed for airtight sample storage. The uniform drop size can be used for reproducible spotting of gels and TLC plates and dispensing into test plates, ID strips, PCR tubes, or 96-well plates. Sterile pipettes are also excellent for tissue culture application.
Catalog No. 50-238-9527 Supplier Alkali Scientific Supplier No. DP412100
These unbreakable, one-piece pipettes eliminate contaminated rubber bulbs, broken glass, and exposure to infectious materials. They are made of ultra-clear, low density polyethylene (LDPE) and are ideal for microbiology, blood banking, hematology, immunology, and urinalysis. Their low-affinity surface reduces sample, cell, or protein loss, and they can also be heat-sealed for airtight sample storage. The uniform drop size can be used for reproducible spotting of gels and TLC plates and dispensing into test plates, ID strips, PCR tubes, or 96-well plates. Sterile pipettes are also excellent for tissue culture application.