Learn More
Andwin Scientific TRANSFER PIPETTE 12ML 50/PK
SDP
Catalog No. 50930644
Encompass_Preferred
The 12 mL transfer pipette is ideal for procedures that requires transfering of liquids. This product is constructed with polypropylene and is able to be attached to tubings to complete desired applications. The tips may also be cut to help in the aspiration and delivery of fluids of different viscosities. This is sold in a pack of 50.
Catalog No. 50-930-644 Supplier Andwin Scientific Supplier No. 7098600
The 12 mL transfer pipette is ideal for procedures that requires transfering of liquids. This product is constructed with polypropylene and is able to be attached to tubings to complete desired applications. The tips may also be cut to help in the aspiration and delivery of fluids of different viscosities. This is sold in a pack of 50.