Learn More
Cole-Parmer Cole-Parmer Transfer Pipette, Jumbo-Bulb, Long-Stem, 15.0 mL; 250/Pk
Catalog No. NC0173812 Shop All Cole-Parmer Products
Encompass_Preferred
Use these unbreakable low-density polyethylene (LDPE) pipettes to eliminate the chance of contamination from rubber bulbs. They deliver repeatable, uniform drops and are configured to fit into a variety of vessels. Choose the bulb draw and graduations or drops/mL needed for measurement and delivery.
Catalog No. NC0173812 Supplier Cole-Parmer Supplier No. 0622651
Use these unbreakable low-density polyethylene (LDPE) pipettes to eliminate the chance of contamination from rubber bulbs. They deliver repeatable, uniform drops and are configured to fit into a variety of vessels. Choose the bulb draw and graduations or drops/mL needed for measurement and delivery.