Learn More
GENTECH SOLUTIONS LLC Transfer Pipette 1mL, Bulk
Catalog No. NC3728796
Encompass_Preferred
EliteChem Simplify your liquid handling with these easy-to-use tools, perfect for safe, mess-free sample transfer in lab and clinical settings. Made from flexible, inert polyethylene, they offer precise control and are ideal for single-use applications to reduce cross-contamination. Available in various sizes, bulb capacities, and sterile options to suit a wide range of tasks.
Catalog No. NC3728796 Supplier GENTECH SOLUTIONS LLC Supplier No. 1800300010
EliteChem Simplify your liquid handling with these easy-to-use tools, perfect for safe, mess-free sample transfer in lab and clinical settings. Made from flexible, inert polyethylene, they offer precise control and are ideal for single-use applications to reduce cross-contamination. Available in various sizes, bulb capacities, and sterile options to suit a wide range of tasks.