Learn More
Research Products International Corp Disposable Plastic Transfer Pipettes, General Purpose Large Bulb, 8.0ml Capacity, 400 per Package
SDP
Catalog No. 50212371
Encompass
Plastic transfer pipettes are molded into one-piece units from low density polyethylene. They eliminate cross-contamination encountered with rubber bulbs and reduce exposure when working with hazardous or infectious samples. Ideal for all laboratory procedures that require fast and safe transferring of liquids. Pipettes can also be used as freeze vials or as storage and transport containers by merely heat sealing the tips closed after filling. They can be sterilized if required by gas or radiation. Pipettes are supplied non-sterile. Large capacity. Fits small standard 10mm test tubes. Ideal for large volume transfers.
Catalog No. 50-212-371 Supplier Research Products International Corp Supplier No. 147504
Plastic transfer pipettes are molded into one-piece units from low density polyethylene. They eliminate cross-contamination encountered with rubber bulbs and reduce exposure when working with hazardous or infectious samples. Ideal for all laboratory procedures that require fast and safe transferring of liquids. Pipettes can also be used as freeze vials or as storage and transport containers by merely heat sealing the tips closed after filling. They can be sterilized if required by gas or radiation. Pipettes are supplied non-sterile. Large capacity. Fits small standard 10mm test tubes. Ideal for large volume transfers.